missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Vinyl Mini Pictograms Hazardous Labels
Brand: Brady™ 814015
9.59 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.00270kg
Specifications
ISO 7010, RoHS compliant using EU Directive 2002/95/EC | |
Vinyl | |
Permanent Acrylic | |
10 mm | |
105 mm | |
Diamond | |
Evacuation Panel Safety Sign |
Gloss | |
96/Pk. | |
Pipe markers and safety signs | |
0.061 mm | |
-18°C to 60°C | |
Inflammable Solid Material | |
Black on White |