missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Vinyl Mini Pictograms Hazardous Labels

Product Code. 12024128
Click to view available options
Displayed Text:
CMR Mutagenic - Respiratory Risk
Combustible Material
Corrosive Material
DANGER for Aquatic Environment
DANGER of Cutaneous Sensitivity - inhalation
Explosive Material
Gases Under Pressure
Inflammable Solid Material
Toxic Material
Unit Size:
Pack of 96
This item is not returnable. View return policy

Product Code. 12024128

Brand: Brady™ 814017

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Certifications/Compliance ISO 7010, RoHS compliant using EU Directive 2002/95/EC
Alternate Part Number MINI PIC1804-10X10-GHS04/96
Material Vinyl
Quantity 96/Pk.
Adhesive Type Permanent Acrylic
For Use With (Application) Pipe markers and safety signs
Height (Metric) 10 mm
Width (Metric) 105 mm
Temperature Range (Metric) -18°C to 60°C
Shape Diamond
Displayed Text Gases Under Pressure
Product Type Evacuation Panel Safety Sign
Color Black on White
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.