missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Vinyl with Polyester Overlaminate Durable Marking Tape

£10.35 - £26.92

Specifications

Material Vinyl
Finish High Gloss
For Use With (Application) Safety marking of obstructions, low hanging objects, pillars, passageways, aisles, and any other potentially hazardous areas or objects
Length (Metric) 16.5 m
Packaging Quantity 1 Roll
View More Specs

Products 15
Product Code Brand Description Dimensions (L x W) Price Quantity & Availability  
Product Code Brand Description Dimensions (L x W) Price Quantity & Availability  
11791605
View Documents
Brady™
055290
Durable Marking Tape - Black/White 16.5 m x 25 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11711615
View Documents
Brady™
055292
Durable Marking Tape - Black/White 16.5 m x 50 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11701615
View Documents
Brady™
055291
Durable Marking Tape - Black/White 16.5 m x 38 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11721615
View Documents
Brady™
055293
Durable Marking Tape - Black/White 16.5 m x 75 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11771615
View Documents
Brady™
055300
Durable Marking Tape - Yellow/Black 16.5 m x 25 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11781615
View Documents
Brady™
055301
Durable Marking Tape - Yellow/Black 16.5 m x 38 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11701625
View Documents
Brady™
055303
Durable Marking Tape - Yellow/Black 16.5 m x 75 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11791615
View Documents
Brady™
055302
Durable Marking Tape - Yellow/Black 16.5 m x 50 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11711625
View Documents
Brady™
055312
Durable Marking Tape - Magenta/Yellow 16.5 m x 38 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11721875
View Documents
Brady™
055313
Durable Marking Tape - Magenta/Yellow 16.5 m x 50 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11731875
View Documents
Brady™
055314
Durable Marking Tape - Magenta/Yellow 16.5 m x 75 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11731615
View Documents
Brady™
055294
Durable Marking Tape - Red/White 16.5 m x 25 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11741615
View Documents
Brady™
055295
Durable Marking Tape - Red/White 16.5 m x 38 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11751615
View Documents
Brady™
055296
Durable Marking Tape - Red/White 16.5 m x 50 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11761615
View Documents
Brady™
055297
Durable Marking Tape - Red/White 16.5 m x 75 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Specifications

Vinyl
Safety marking of obstructions, low hanging objects, pillars, passageways, aisles, and any other potentially hazardous areas or objects
1 Roll
0.037 mm
High Gloss
16.5 m
-40°C to +80°C
Videos
SDS
Documents

Documents

Product Certifications