missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Vinyl Pre-Printed Chemical Labels
Brady™ Vinyl GHS/CLP Chemical Labels for BBP3x printers are self-adhesive and preprinted with red diamonds in a variety of layouts.
£370.00
Specifications
Color | White, Red |
---|---|
Finish | Gloss |
Material | Vinyl |
Product Type | Pre-printed Chemical Label |
For Use With (Application) | Facility and Safety identification, Hazardous Substance identification |
Product Code | Brand | Description | Height (Metric) | Width (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Height (Metric) | Width (Metric) | Price | Quantity & Availability | |||
15326651
|
Brady™
121647 |
3 Diamonds | 155 mm | 105 mm |
£370.00
1 roll |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13573340
|
Brady™
121646 |
2 Diamonds, vertical | 155.00 mm | 105 mm |
£370.00
1 roll |
In Stock
Log in to see stock. |
|||
13543340
|
Brady™
121645 |
1 Diamond | 155.00 mm | 105 mm |
£370.00
1 roll |
In Stock
Log in to see stock. |
|||
Description
- BBP™31 and BBP™33 compatible labels
- Blank labels with pre-printed red diamonds allow printer to produce two-color GHS/CLP labels in one print pass
- Available in three sizes in the full product portfolio, each with six diamond layouts that match templates in MarkWare™Software
- Service temperature -20°C to 75°C; minimum application temperature 10°C
- Average outdoor durability 3-5 years
Specifications
White, Red | |
Vinyl | |
Facility and Safety identification, Hazardous Substance identification | |
MarkWare | |
105 mm |
Gloss | |
Pre-printed Chemical Label | |
Up to 3 Years Outdoor Use | |
-20°C to +75°C |