missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Allsigns International Safety sign WA144/SA 200mm x 300mm hazard warning self adhesive vinyl
Brand: Allsigns International WA145S
Safety sign WA144/SA 200mm x 300mm hazard warning self adhesive vinyl
Code : 04
Additional Details : Weight : 0.01504kg