missing translation for 'onlineSavingsMsg'
Learn More

3M™ Vinyl Tape 471F

Product Code. 17277770
Click to view available options
Color:
Black
Blue
Green
Red
White
Yellow
Unit Size:
24 rolls
This item is not returnable. View return policy

Product Code. 17277770

Brand: 3M™ 7000028852

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Commonly used for marking hazards and aisles, color-coding, paint masking, decorative trim, splicing, and more. Made from vinyl backing with rubber adhesive, our versatile tape resists scrapes, wear, and weathering for long lasting use.

  • Maintains its color when exposed to abrasion, wear and many solvents
  • Unique stretch properties enable tape to remain stretched to conform to uneven surfaces without lifting and retracting
  • Ideal for use on curved and irregular surfaces
  • Easy to apply, excellent holding strength on many surfaces
  • Available in many distinctive colours (yellow, white, red, black, brown, green, orange, purple, blue and transparent)
  • Available in a wide variety of vivid colours and sizes
TRUSTED_SUSTAINABILITY

Specifications

Dimensions (L x W) 50 mm x 33 m
Height (Metric) 0.1 mm
Length (Metric) 32.9 m
Thickness 0.14 mm
Width (Metric) 5.1 cm
Color Yellow
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.