Learn More
Vinylcyclopentane, 99%, Thermo Scientific Chemicals
£53.16 - £229.14
Chemical Identifiers
CAS | 3742-34-5 |
---|---|
Molecular Formula | C7H12 |
Molecular Weight (g/mol) | 96.17 |
MDL Number | MFCD00001386 |
InChI Key | BEFDCLMNVWHSGT-UHFFFAOYSA-N |
Synonym | vinylcyclopentane, cyclopentane, ethenyl, cyclopentane, vinyl, cyclopentyl-ethene, cyclopentylethylene, cyclopentane,ethenyl, acmc-209iss |
PubChem CID | 77345 |
IUPAC Name | ethenylcyclopentane |
SMILES | C=CC1CCCC1 |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3742-34-5 | |
96.17 | |
BEFDCLMNVWHSGT-UHFFFAOYSA-N | |
77345 | |
C=CC1CCCC1 |
C7H12 | |
MFCD00001386 | |
vinylcyclopentane, cyclopentane, ethenyl, cyclopentane, vinyl, cyclopentyl-ethene, cyclopentylethylene, cyclopentane,ethenyl, acmc-209iss | |
ethenylcyclopentane |
Specifications
3742-34-5 | |
97°C | |
C7H12 | |
MFCD00001386 | |
vinylcyclopentane, cyclopentane, ethenyl, cyclopentane, vinyl, cyclopentyl-ethene, cyclopentylethylene, cyclopentane,ethenyl, acmc-209iss | |
C=CC1CCCC1 | |
96.17 | |
96.17 | |
Vinylcyclopentane |
0.704 | |
10°C (50°F) | |
1.436 | |
UN3295 | |
BEFDCLMNVWHSGT-UHFFFAOYSA-N | |
ethenylcyclopentane | |
77345 | |
99% |
Safety and Handling
GHS H Statement
H225
Highly flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H225
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
missing translation for 'einecsNumber' : 223-133-8
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only