Learn More
Vinylene carbonate, 98%, stabilized, Thermo Scientific Chemicals
£43.30 - £145.00
Chemical Identifiers
CAS | 872-36-6 |
---|---|
Molecular Formula | C3H2O3 |
Molecular Weight (g/mol) | 86.05 |
MDL Number | MFCD00005380 |
InChI Key | VAYTZRYEBVHVLE-UHFFFAOYSA-N |
Synonym | vinylene carbonate, vinyl carbonate, carbonic acid, cyclic vinylene ester, wln: t5ovoj, vinylenecarbonate, vinyleny carbonate, carbonic acid vinylene, 1,3-dioxo-2-one, vinylene carbonate vc, acmc-209qj3 |
PubChem CID | 13385 |
SMILES | O=C1OC=CO1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10068571
|
Thermo Scientific
140890050 |
5 g | Glass bottle |
£43.30
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10611073
|
Thermo Scientific Acros
140890250 |
25 g | Glass bottle |
£145.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Chemical Identifiers
872-36-6 | |
86.05 | |
VAYTZRYEBVHVLE-UHFFFAOYSA-N | |
13385 |
C3H2O3 | |
MFCD00005380 | |
vinylene carbonate, vinyl carbonate, carbonic acid, cyclic vinylene ester, wln: t5ovoj, vinylenecarbonate, vinyleny carbonate, carbonic acid vinylene, 1,3-dioxo-2-one, vinylene carbonate vc, acmc-209qj3 | |
O=C1OC=CO1 |
Specifications
872-36-6 | |
Colorless to Yellow | |
162°C | |
Authentic | |
C3H2O3 | |
MFCD00005380 | |
13,342 | |
vinylene carbonate, vinyl carbonate, carbonic acid, cyclic vinylene ester, wln: t5ovoj, vinylenecarbonate, vinyleny carbonate, carbonic acid vinylene, 1,3-dioxo-2-one, vinylene carbonate vc, acmc-209qj3 | |
VAYTZRYEBVHVLE-UHFFFAOYSA-N | |
2H-1,3-dioxol-2-one | |
86.05 | |
86.05 | |
Liquid |
19°C to 22°C | |
1.3600g/mL | |
73°C | |
97.5% min. (GC) | |
1.4200 to 1.423 | |
19, V,4, 72 | |
1.36 | |
Solubility in water: 11.5g / 100mL | |
O=C1OC=CO1 | |
0.1% max. BHT | |
13385 | |
98% | |
Vinylene carbonate, Stabilized |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye damage.
May cause damage to organs through prolonged or repeated exposu
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minut
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 212-825-5
- Vinylene carbonate, stabilized
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity after repeated exposure Category 2
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H302-Harmful if swallowed.
- H311-Toxic in contact with skin.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap and water.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/physician.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
RUO – Research Use Only