Learn More
Vinylidene chloride, 99.9%, stabilized, Thermo Scientific Chemicals
£63.40 - £144.00
Chemical Identifiers
CAS | 75-35-4, 150-76-5 |
---|---|
MDL Number | MFCD00011653 |
InChI Key | LGXVIGDEPROXKC-UHFFFAOYSA-N |
Synonym | vinylidene chloride, 1,1-dichloroethylene, vinylidene dichloride, 1,1-dce, ethene, 1,1-dichloro, vinylidine chloride, ethylene, 1,1-dichloro, as-dichloroethylene, rcra waste number u078, asym-dichloroethylene |
PubChem CID | 6366 |
ChEBI | CHEBI:34031 |
IUPAC Name | 1,1-dichloroethene |
SMILES | C=C(Cl)Cl |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
75-35-4, 150-76-5 | |
LGXVIGDEPROXKC-UHFFFAOYSA-N | |
6366 | |
1,1-dichloroethene |
MFCD00011653 | |
vinylidene chloride, 1,1-dichloroethylene, vinylidene dichloride, 1,1-dce, ethene, 1,1-dichloro, vinylidine chloride, ethylene, 1,1-dichloro, as-dichloroethylene, rcra waste number u078, asym-dichloroethylene | |
CHEBI:34031 | |
C=C(Cl)Cl |
Specifications
75-35-4, 150-76-5 | |
Colorless | |
31.2°C to 31.7°C | |
Authentic | |
C2H2Cl2 | |
1.4244 to 1.4264 | |
01, 186 | |
1.218 | |
LGXVIGDEPROXKC-UHFFFAOYSA-N | |
1,1-dichloroethene | |
96.938 | |
CHEBI:34031 | |
99.9% | |
Vinylidene chloride, 99.9% |
-122.0°C | |
1.2180g/mL | |
−25°C | |
99.90% | |
H2C=CCl2 | |
MFCD00011653 | |
15, 10192 | |
vinylidene chloride, 1,1-dichloroethylene, vinylidene dichloride, 1,1-dce, ethene, 1,1-dichloro, vinylidine chloride, ethylene, 1,1-dichloro, as-dichloroethylene, rcra waste number u078, asym-dichloroethylene | |
C=C(Cl)Cl | |
180 to 220ppm MEHQ | |
6366 | |
96.94 | |
Liquid |
Safety and Handling
GHS H Statement:
Suspected of causing cancer.
Harmful if inhaled.
Extremely flammable liquid and vapor.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.
Store in a well-ventilated place. Keep cool.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 200-864-0
missing translation for 'rtecsNumber' : KV9275000
missing translation for 'tsca' : TSCA
- Vinylidene chloride, stabilized
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 1
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H351-Suspected of causing cancer.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H224-Extremely flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P243-Take precautionary measures against static discharge.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P403+P235-Store in a well-ventilated place. Keep cool.
RUO – Research Use Only