Learn More
Vinyltrichlorosilane, 97%, Thermo Scientific Chemicals
£27.44 - £76.48
Chemical Identifiers
CAS | 75-94-5 |
---|---|
Molecular Formula | C2H3Cl3Si |
Molecular Weight (g/mol) | 161.481 |
MDL Number | MFCD00000482 |
InChI Key | GQIUQDDJKHLHTB-UHFFFAOYSA-N |
Synonym | trichlorovinylsilane, vinyltrichlorosilane, trichloro vinyl silane, vtcs, silane, trichloroethenyl, silane, trichlorovinyl, trichlorovinylsilicon, vinyl trichlorosilane, trichlorovinyl silicane, vinylsilicon trichlorideShow More |
PubChem CID | 6413 |
IUPAC Name | trichloro(ethenyl)silane |
SMILES | C=C[Si](Cl)(Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11346926
|
Thermo Scientific Alfa Aesar
L04387.22 |
100 g | N/A | ||||||
11351567
|
Thermo Scientific Alfa Aesar
L04387.36 |
500 g |
£76.48
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
75-94-5 | |
161.481 | |
GQIUQDDJKHLHTB-UHFFFAOYSA-N | |
6413 | |
C=C[Si](Cl)(Cl)Cl |
C2H3Cl3Si | |
MFCD00000482 | |
trichlorovinylsilane, vinyltrichlorosilane, trichloro vinyl silane, vtcs, silane, trichloroethenyl, silane, trichlorovinyl, trichlorovinylsilicon, vinyl trichlorosilane, trichlorovinyl silicane, vinylsilicon trichlorideShow More | |
trichloro(ethenyl)silane |
Specifications
91°C to 93°C | |
75-94-5 | |
CH2=CHSiCl3 | |
UN1305 | |
10°C (50°F) | |
GQIUQDDJKHLHTB-UHFFFAOYSA-N | |
trichloro(ethenyl)silane | |
6413 | |
97% | |
Moisture sensitive | |
Vinyltrichlorosilane |
-95°C | |
C2H3Cl3Si | |
MFCD00000482 | |
1743440 | |
trichlorovinylsilane, vinyltrichlorosilane, trichloro vinyl silane, vtcs, silane, trichloroethenyl, silane, trichlorovinyl, trichlorovinylsilicon, vinyl trichlorosilane, trichlorovinyl silicane, vinylsilicon trichloride | |
C=C[Si](Cl)(Cl)Cl | |
161.481 | |
161.49 | |
1.436 | |
1.27 |
Safety and Handling
GHS H Statement
H225-H311-H314-H318-H302-H332
Highly flammable liquid and vapor.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H225-H302+H332-H311-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: VINYLTRICHLOROSILANE
missing translation for 'einecsNumber' : 200-917-8
missing translation for 'rtecsNumber' : VV6125000
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.