missing translation for 'onlineSavingsMsg'
Learn More
Vinyltriethoxysilane, 97%, Thermo Scientific Chemicals Product Code.: 11400137

Vinyltriethoxysilane, 97%, Thermo Scientific Chemicals

Product Code. 11400137
250 g
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy

Product Code. 11400137

Brand: Thermo Scientific Alfa Aesar B21037.30

This item is not returnable. View return policy

Vinyltriethoxysilane is used as an adhesive for glass slides. It acts as a coupling agent on inorganic substances blended into polymers that function as adhesion promoters, crosslinkers, and surface modifiers. Further, it is used to prepare polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Vinyltriethoxysilane is used as an adhesive for glass slides. It acts as a coupling agent on inorganic substances blended into polymers that function as adhesion promoters, crosslinkers, and surface modifiers. Further, it is used to prepare polymers.

Solubility
Miscible with organic solvents.

Notes
Moisture sensitive. Incompatible with alkali, acid, water and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-08-0
Molecular Formula C8H18O3Si
Molecular Weight (g/mol) 190.31
MDL Number MFCD00009063
InChI Key FWDBOZPQNFPOLF-UHFFFAOYSA-N
Synonym vinyltriethoxysilane, triethoxyvinylsilane, silane, ethenyltriethoxy, triethoxy vinyl silane, polyscience vtes, triethoxyvinylsilicane, vtes, silane a 151, ethenyl triethoxy silane, silane, triethoxyvinylShow MoreShow Less
PubChem CID 6516
IUPAC Name ethenyl(triethoxy)silane
SMILES CCO[Si](OCC)(OCC)C=C

Specifications

Density 0.903
Boiling Point 160°C to 161°C
Flash Point 34°C (93°F)
Assay Percent Range 0.97
Odor Fruit-like
Linear Formula (CH3CH2O)3SiCH=CH2
Refractive Index 1.399
Quantity 250 g
UN Number UN1993
Beilstein 1767229
Sensitivity Moisture sensitive
Solubility Information Miscible with organic solvents.
Formula Weight 190.31
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Vinyltriethoxysilane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252032 Certificate TypeCertificate of Analysis Date13/08/2024 Catalog NumberB21037.18,B21037.30,B21037.0B
Lot Number10248608 Certificate TypeCertificate of Analysis Date08/11/2023 Catalog NumberB21037.18,B21037.30,B21037.0B
Lot Number10188135 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21037.18,B21037.30,B21037.0B
Lot Number10168571 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21037.18,B21037.30,B21037.0B
Lot NumberUSLF003545 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21037.18,B21037.30,B21037.0B

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Vinyltriethoxysilane, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.