missing translation for 'onlineSavingsMsg'
Learn More

Vinyltrimethoxysilane, 98%, Thermo Scientific Chemicals

Product Code. 10570804
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2768-02-7
Molecular Formula C5H12O3Si
Molecular Weight (g/mol) 148.24
MDL Number MFCD00008605
InChI Key NKSJNEHGWDZZQF-UHFFFAOYSA-N
Synonym vinyltrimethoxysilane, trimethoxyvinylsilane, silane, ethenyltrimethoxy, trimethoxy vinyl silane, trimethoxysilyl ethene, vinyl trimethoxy silane, silane, trimethoxyvinyl, trimethoxysilyl ethylene, vtmo, vts-m
PubChem CID 76004
IUPAC Name ethenyl(trimethoxy)silane
SMILES CO[Si](C=C)(OC)OC

Specifications

Density 0.9680g/mL
Boiling Point 123.0°C
Flash Point 23°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHSi(OCH3)3
Refractive Index 1.3915 to 1.3935
Quantity 500 g
Specific Gravity 0.968
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in most common organic solvents
Viscosity 0.6 mm2/s (25°C)
Formula Weight 148.24
Percent Purity 98%
Chemical Name or Material Vinyltrimethoxysilane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Vinyltrimethoxysilane
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
  • Skin sensitiser Category 1
Hazard Statement
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P362+P364-Take off contaminated clothing and wash it before reuse.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'