missing translation for 'onlineSavingsMsg'
Learn More

Vinyltrimethylsilane, 97%, Thermo Scientific Chemicals

Product Code. 10635272
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 754-05-2
Molecular Formula C5H12Si
Molecular Weight (g/mol) 100.24
MDL Number MFCD00008606
InChI Key GCSJLQSCSDMKTP-UHFFFAOYSA-N
Synonym vinyltrimethylsilane, trimethyl vinyl silane, trimethylvinylsilane, silane, ethenyltrimethyl, trimethylsilyl ethylene, vinyl trimethylsilane, silane, trimethylvinyl, vinyltrimethysilane, trimethylsilyl ethene, ethenyl-trimethylsilane
PubChem CID 79102
IUPAC Name ethenyl(trimethyl)silane
SMILES C[Si](C)(C)C=C

Specifications

Color Colorless
Density 0.6800g/mL
Boiling Point 55°C
Flash Point -24°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHSi(CH3)3
Refractive Index 1.3895 to 1.3915
Quantity 25 mL
Fieser 09,498; 10,444; 12,566; 13,343
Specific Gravity 0.68
Formula Weight 100.24
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Vinyltrimethylsilane, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Vinyltrimethylsilane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'