Learn More
Vinyltrimethylsilane, 97%, Thermo Scientific Chemicals
£36.88 - £480.00
Chemical Identifiers
CAS | 754-05-2 |
---|---|
Molecular Formula | C5H12Si |
Molecular Weight (g/mol) | 100.236 |
MDL Number | MFCD00008606 |
InChI Key | GCSJLQSCSDMKTP-UHFFFAOYSA-N |
Synonym | vinyltrimethylsilane, trimethyl vinyl silane, trimethylvinylsilane, silane, ethenyltrimethyl, trimethylsilyl ethylene, vinyl trimethylsilane, silane, trimethylvinyl, vinyltrimethysilane, trimethylsilyl ethene, ethenyl-trimethylsilane |
PubChem CID | 79102 |
IUPAC Name | ethenyl(trimethyl)silane |
SMILES | C[Si](C)(C)C=C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11347725
|
Thermo Scientific Alfa Aesar
L02498.09 |
10 g |
£36.88
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11357725
|
Thermo Scientific Alfa Aesar
L02498.18 |
50 g |
£122.61
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11367725
|
Thermo Scientific Alfa Aesar
L02498.30 |
250 g |
£480.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Vinyltrimethylsilane is used in semiconductor processing. It is involved in the Preparation of silyl-ethers by Rh(I) catalysis. Further, it is used in the preparation of trimethylsilylethyl sulfoxides which in the presence of fluoride ion generate sulfenate nucleophiles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsVinyltrimethylsilane is used in semiconductor processing. It is involved in the Preparation of silyl-ethers by Rh(I) catalysis. Further, it is used in the preparation of trimethylsilylethyl sulfoxides which in the presence of fluoride ion generate sulfenate nucleophiles.
Solubility
Miscible with tetrahydrofuran, diethyl ether, benzene and dichloromethane.
Notes
Store in a cool place. Hygroscopic. Incompatible with strong oxidizing agents, strong acids and strong alkalis (bases).
Chemical Identifiers
754-05-2 | |
100.236 | |
GCSJLQSCSDMKTP-UHFFFAOYSA-N | |
79102 | |
C[Si](C)(C)C=C |
C5H12Si | |
MFCD00008606 | |
vinyltrimethylsilane, trimethyl vinyl silane, trimethylvinylsilane, silane, ethenyltrimethyl, trimethylsilyl ethylene, vinyl trimethylsilane, silane, trimethylvinyl, vinyltrimethysilane, trimethylsilyl ethene, ethenyl-trimethylsilane | |
ethenyl(trimethyl)silane |
Specifications
54°C to 56°C | |
754-05-2 | |
MFCD00008606 | |
956572 | |
vinyltrimethylsilane, trimethyl vinyl silane, trimethylvinylsilane, silane, ethenyltrimethyl, trimethylsilyl ethylene, vinyl trimethylsilane, silane, trimethylvinyl, vinyltrimethysilane, trimethylsilyl ethene, ethenyl-trimethylsilane | |
GCSJLQSCSDMKTP-UHFFFAOYSA-N | |
ethenyl(trimethyl)silane | |
79102 | |
97% | |
1.391 | |
Vinyltrimethylsilane |
-132°C | |
C5H12Si | |
UN1993 | |
−24°C (−11°F) | |
Miscible with tetrahydrofuran,diethyl ether,benzene and dichloromethane. | |
C[Si](C)(C)C=C | |
100.236 | |
100.24 | |
Mild | |
0.69 |
Safety and Handling
GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'einecsNumber' : 212-042-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only