missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Violanthrone-79, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
Description
Violanthrone 79 is an n-channel organic semiconductor used for interface control of conventional n-type meta diode.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Violanthrone 79 is an n-channel organic semiconductor used for interface control of conventional n-type meta diode.
Solubility
Soluble in chloroform.
Notes
Refrigerated. Incompatible with strong oxi
Violanthrone 79 is an n-channel organic semiconductor used for interface control of conventional n-type meta diode.
Solubility
Soluble in chloroform.
Notes
Refrigerated. Incompatible with strong oxi
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
CAS | 85652-50-2 |
Melting Point | 193°C to 196°C |
Molecular Formula | C50H48O4 |
MDL Number | MFCD00491570 |
Quantity | 1 g |
Synonym | violanthrone-79, violanthrone 79, anthra 9,1,2-cde benzo rst pentaphene-5,10-dione,16,17-bis octyloxy, anthra 9,1,2-cde benzo rst pentaphene-5,10-dione,1, 16,17-bis n-octyloxy anthra 9,1,2-cde benzo rst pentaphene-5,10-dione, 16,17-bis octyloxy-anthra 9,1,2-cde benzo rst pentaphene-5,10-dione, 16,17-bis octyloxy anthra 9,1,2-cde-benzo rst pentaphene-5,10-dione, 16,17-bis octyloxy anthra 9,1,2-cde benzo rst pentaphene-5,10-dione |
Solubility Information | Soluble in chloroform. |
InChI Key | LLPQZABTDLOYAL-UHFFFAOYSA-N |
SMILES | CCCCCCCCOC1=C2C3=C(C=CC4=C3C(=C1)C5=CC=CC=C5C4=O)C6=C7C2=C(C=C8C7=C(C=C6)C(=O)C9=CC=CC=C98)OCCCCCCCC |
Molecular Weight (g/mol) | 712.93 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'