missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Violet Ratiometric Membrane Asymmetry Probe/Dead Cell Apoptosis Kit, for flow cytometry

Violet Ratiometric Membrane Asymmetry Probe/Dead Cell Apoptosis Kit, for flow cytometry

Brand:  Molecular Probes™ A35137

Product Code. 10630930

  • £437.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

The Violet Ratiometric Membrane Asymmetry Probe/Dead Cell Apoptosis Kit provides a simple and fast method for the detection of apoptosis with dead cell discrimination by flow cytometry. The Violet Ratiometric Membrane Asymmetry Probe, 4'-N,N-diethylamino-6-(N,N,N-dodecyl-methylamino-sulfopropyl)-methyl-3-hydroxyflavone (F2N12S), is a novel violet excitable dye for the detection of membrane asymmetry changes during apoptosis. The dye exhibits an excited-state intramolecular proton transfer (ESIPT) reaction resulting in a dual fluorescence with two emission bands corresponding to 530 nm and 585 nm, producing a two-color ratiometric response to variations in surface charge. Ratiometric probes have several advantages over traditional fluorochrome labeled reagents. The ratiometric probe is a self-calibrating absolute parameter of apoptotic transformation, which is independent of probe concentration, cell size, and instrument variation, such as fluctuations of laser intensity or sensitivity of the detectors. Given that apoptosis modifies the surface charge of the outer leaflet of the plasma membrane the violet membrane asymmetry probe F2N12S can monitor changes in membrane asymmetry that occur during apoptosis through a change in the relative intensity of the two emission bands of the dye. The F2N12S probe is combined with SYTOX(R) AADvanced dead cell stain, which is capable of passing through the cell membrane only in late apoptotic or necrotic cells allowing discrimination form early apoptotic cells. Samples can be analyzed after a 5 minute incubation at room temperature and does not require special buffers or wash steps. This kit can be paired with other reagents such as MitoProbe™ DiIC1(5) or annexin V for multiparametric analysis of apoptosis and viability. This reagent kit allow researchers to maximaize the utility of their instruments by utilizing the violet laser Each kit contains sufficient reagents for ∼100 flow cytometry tests.

View a selection guide for all apoptosis assays for flow cytometry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 kit
Dead Cell Apoptosis Kit
405, 488
100
Fluorescence
Flow Cytometer
Other Label(s) or Dye(s)
Tube
Contains 1 vial of F2N12S (100 μL), 1 vial of SYTOX™ AADvanced™ dead cell stain, and 1 vial of DMSO (200 μL).

Store at -20°C, dessicate and protect from light.
SYTOX™ AADvanced™: 546/647
F2N12S: 405/530
Dry Ice
Flow Cytometry
F2N12S, SYTOX AADvanced Dead Cell Stain
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.