missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Viral Production Medium

Product Code. 17371889
Click to view available options
Quantity:
1 L
6 L
10 L
Unit Size:
1 L
10 L
Pack of 6
This item is not returnable. View return policy

Product Code. 17371889

missing translation for 'mfr': Gibco™ A4817901

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Viral Production Medium is a chemically defined, serum-free, and protein-free medium for growth and transfection of Viral Production Cells 2.0.

Gibco Viral Production Medium is a chemically defined, serum-free, and protein-free medium. It is animal origin-free and specially developed for growth and transfection of Viral Production Cells 2.0 (suspension-adapted HEK 293F-derived cells).

  • For adeno-associated virus (AAV) production as part of the AAV-MAX Helper-Free AAV Production System
  • Supports growth of suspension Viral Production Cells 2.0 to densities of over 1.2 x 107 cells/mL
  • Enables production of high AAV titers, greater than 5 x 1010 vg/mL (unpurified)
  • Supports growth and transfection of Viral Production Cells 2.0 from shake flask to bioreactor scales
  • Viral Production Medium requires supplementation with 4 mM GlutaMAX Supplement (Cat. No. 35050038) prior to use
TRUSTED_SUSTAINABILITY

Tekniske data

Cell Line Viral Production Cells 2.0
Cell Type HEK 293
Classification Serum-free, Animal Origin-free, Chemically-defined, Protein-free
Form Liquid
Product Type AAV Production Medium
Sterility Sterile-filtered
With Additives Glucose, Sodium Pyruvate, Sodium Pyruvate, GlutaMAX
Without Additives No GlutaMAX
For Use With (Application) AAV Production
Manufacturing Quality ISO 13485, MDSAP, FDA-registered
Product Line AAV-MAX
Quantity 1 L
Shipping Condition Room Temperature
Format Bottle
pH 7
Regulatory Status RUO - Research Use Only
Content And Storage 1 L bottle; Reagents should be stored at 2°C to 8°C, protected from light
Culture Type Suspension Cell Culture
Expression System Mammalian
Vis mere Vis mindre

For Research Use Only. Not for use in diagnostic procedures.

Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.