Learn More
Viral Production Medium, prototype
Viral Production Medium, prototype, requires supplementation with 4 mM GlutaMAX Supplement prior to use.
Brand:  Invitrogen A49842DK
Additional Details : Weight : 1.11000kg
Product Code. 17133259
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Gibco Viral Production Medium, prototype, is a chemically defined, serum-free, protein-free medium specially developed for growth and transfection of Viral Production Cells 2.0 (suspension-adapted HEK 293F-derived cells) for adeno-associated virus (AAV) production as part of the Gibco AAV-MAX Helper-Free AAV production system.
Specifications
Specifications
Store at 2–8°C, protected from light.
Mammalian Cells
Mammalian
GlutaMAX
AAV Production
RUO
HEK293
Suspension Cell Culture
Liquid
Bottle
AAV-MAX
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Research Use Only. Not for use in diagnostic procedures. This product is a prototype and performance characteristics of this product have not been established.