missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ ViralSEQ™ Mouse Minute Virus (MMV) Detection System

The Applied Biosystems ViralSEQ Mouse Minute Virus (MMV) Detection Assay is part of an integrated DNA purification and real-time PCR-based system for rapid, sensitive detection of MMV from a wide range of sample types.

Brand:  Applied Biosystems™ 4444415

Product Code. 30000690

  • £3165.00 / 100 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Contains Real-Time PCR assay with buffer and enzyme mix, positive control and negative controls
High
Standard
Mouse Minute Virus Detection System
≥ 10 copies MMV DNA
Mouse
Target-Specific
Viral DNA
Primer-probe
qPCR
ViralSEQ™
100 reactions
Dry Ice
Mouse Minute Virus (MMV)
100 Reactions
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2503069 Certificate TypeCertificate of Analysis Date17/04/2025 Catalog Number
Lot Number2502068 Certificate TypeCertificate of Analysis Date14/03/2025 Catalog Number
Lot Number2410066 Certificate TypeCertificate of Analysis Date03/12/2024 Catalog Number4444415
Lot Number2407065 Certificate TypeCertificate of Analysis Date13/08/2024 Catalog Number4444415
Lot Number2405064 Certificate TypeCertificate of Analysis Date07/06/2024 Catalog Number4444415
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Applied Biosystems™ ViralSEQ™ Mouse Minute Virus (MMV) Detection System > 100 Reactions

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.