Learn More
Waters Corporation Viridis BEH Column
Designed and tested for SFC, the Viridis columns demonstrate a wide range of selectivity for greater resolution.
£964.50 - £25395.00
Specifications
Max. Pressure | 6000 psi (415 Bar) |
---|---|
Particle Shape | Spherical |
Pore Size | 130 Å |
Quantity | 1/Ea. |
Endcapped | No |
Product Code | Brand | Length (Metric) | Diameter (Metric) | Particle Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) | Particle Size | Price | Quantity & Availability | ||||
15229046
![]() |
Waters Corporation
186006558 |
50 mm | 2.1 mm | 1.7 μm |
£1149.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15209016
![]() |
Waters Corporation
186005737 |
100 mm | 30 mm | 5 μm |
£8686.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15695295
![]() |
Waters Corporation
186006640 |
100 mm | 3 mm | 3.5 μm |
£1143.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17123262
![]() |
Waters Corporation
186005719 |
50 mm | 3 mm | 5 μm |
£964.50
Each |
Estimated Shipment: 25-04-2025
Log in to see stock. |
||||
17133262
![]() |
Waters Corporation
186005720 |
100 mm | 3 mm | 5 μm |
£1057.50
Each |
Estimated Shipment: 25-04-2025
Log in to see stock. |
||||
17143262
![]() |
Waters Corporation
186005721 |
150 mm | 3 mm | 5 μm |
£1132.50
Each |
Estimated Shipment: 25-04-2025
Log in to see stock. |
||||
17153262
![]() |
Waters Corporation
186005722 |
250 mm | 3 mm | 5 μm |
£1240.50
Each |
Estimated Shipment: 25-04-2025
Log in to see stock. |
||||
17163262
![]() |
Waters Corporation
186005723 |
50 mm | 4.6 mm | 5 μm |
£982.50
Each |
Estimated Shipment: 25-04-2025
Log in to see stock. |
||||
17173262
![]() |
Waters Corporation
186005724 |
100 mm | 4.6 mm | 5 μm |
£1074.00
Each |
Estimated Shipment: 25-04-2025
Log in to see stock. |
||||
17183262
![]() |
Waters Corporation
186005725 |
150 mm | 4.6 mm | 5 μm |
£1143.00
Each |
Estimated Shipment: 25-04-2025
Log in to see stock. |
||||
17193262
![]() |
Waters Corporation
186005726 |
250 mm | 4.6 mm | 5 μm |
£1252.50
Each |
Estimated Shipment: 25-04-2025
Log in to see stock. |
||||
17103272
![]() |
Waters Corporation
186005731 |
50 mm | 19 mm | 5 μm |
£2700.00
Each |
Estimated Shipment: 25-04-2025
Log in to see stock. |
||||
17113272
![]() |
Waters Corporation
186005732 |
100 mm | 19 mm | 5 μm |
£4263.00
Each |
Estimated Shipment: 25-04-2025
Log in to see stock. |
||||
17123272
![]() |
Waters Corporation
186005733 |
150 mm | 19 mm | 5 μm |
£5355.00
Each |
Estimated Shipment: 25-04-2025
Log in to see stock. |
||||
17133272
![]() |
Waters Corporation
186005734 |
250 mm | 19 mm | 5 μm |
£6021.00
Each |
Estimated Shipment: 25-04-2025
Log in to see stock. |
||||
17143272
![]() |
Waters Corporation
186005735 |
50 mm | 30 mm | 5 μm |
£5814.00
Each |
Estimated Shipment: 25-04-2025
Log in to see stock. |
||||
17153272
![]() |
Waters Corporation
186005736 |
75 mm | 30 mm | 5 μm |
£7215.00
Each |
Estimated Shipment: 25-04-2025
Log in to see stock. |
||||
17163272
![]() |
Waters Corporation
186005738 |
150 mm | 30 mm | 5 μm |
£11835.00
Each |
Estimated Shipment: 25-04-2025
Log in to see stock. |
||||
17173272
![]() |
Waters Corporation
186005739 |
250 mm | 30 mm | 5 μm |
£18615.00
Each |
Estimated Shipment: 25-04-2025
Log in to see stock. |
||||
17183272
![]() |
Waters Corporation
186005740 |
50 mm | 50 mm | 5 μm |
£10548.00
Each |
Estimated Shipment: 25-04-2025
Log in to see stock. |
||||
17193272
![]() |
Waters Corporation
186005741 |
100 mm | 50 mm | 5 μm |
£14478.00
Each |
Estimated Shipment: 25-04-2025
Log in to see stock. |
||||
17103282
![]() |
Waters Corporation
186005742 |
150 mm | 50 mm | 5 μm |
£19650.00
Each |
Estimated Shipment: 25-04-2025
Log in to see stock. |
||||
17113282
![]() |
Waters Corporation
186005743 |
250 mm | 50 mm | 5 μm |
£25395.00
Each |
Estimated Shipment: 25-04-2025
Log in to see stock. |
||||
17113762
![]() |
Waters Corporation
186006544 |
150 mm | 2.1 mm | 5 μm |
£1132.50
Each |
Estimated Shipment: 25-04-2025
Log in to see stock. |
||||
17143762
![]() |
Waters Corporation
186006559 |
75 mm | 2.1 mm | 1.7 μm |
£1183.50
Each |
Estimated Shipment: 25-04-2025
Log in to see stock. |
||||
17153762
![]() |
Waters Corporation
186006560 |
100 mm | 2.1 mm | 1.7 μm |
£1230.00
Each |
Estimated Shipment: 25-04-2025
Log in to see stock. |
||||
17163762
![]() |
Waters Corporation
186006561 |
150 mm | 2.1 mm | 1.7 μm |
£1309.50
Each |
Estimated Shipment: 25-04-2025
Log in to see stock. |
||||
17173762
![]() |
Waters Corporation
186006562 |
50 mm | 3 mm | 1.7 μm |
£1137.00
Each |
Estimated Shipment: 25-04-2025
Log in to see stock. |
||||
17183762
![]() |
Waters Corporation
186006563 |
75 mm | 3 mm | 1.7 μm |
£1183.50
Each |
Estimated Shipment: 25-04-2025
Log in to see stock. |
||||
17193762
![]() |
Waters Corporation
186006564 |
100 mm | 3 mm | 1.7 μm |
£1230.00
Each |
Estimated Shipment: 25-04-2025
Log in to see stock. |
||||
17123822
![]() |
Waters Corporation
186006634 |
50 mm | 2.1 mm | 3.5 μm |
£1102.50
Each |
Estimated Shipment: 25-04-2025
Log in to see stock. |
||||
17133822
![]() |
Waters Corporation
186006635 |
75 mm | 2.1 mm | 3.5 μm |
£1120.50
Each |
Estimated Shipment: 25-04-2025
Log in to see stock. |
||||
17143822
![]() |
Waters Corporation
186006636 |
100 mm | 2.1 mm | 3.5 μm |
£1143.00
Each |
Estimated Shipment: 25-04-2025
Log in to see stock. |
||||
17153822
![]() |
Waters Corporation
186006637 |
150 mm | 2.1 mm | 3.5 μm |
£1206.00
Each |
Estimated Shipment: 25-04-2025
Log in to see stock. |
||||
17163822
![]() |
Waters Corporation
186006638 |
50 mm | 3 mm | 3.5 μm |
£1102.50
Each |
Estimated Shipment: 25-04-2025
Log in to see stock. |
||||
17173822
![]() |
Waters Corporation
186006639 |
75 mm | 3 mm | 3.5 μm |
£1120.50
Each |
Estimated Shipment: 25-04-2025
Log in to see stock. |
||||
17183822
![]() |
Waters Corporation
186006641 |
150 mm | 3 mm | 3.5 μm |
£1206.00
Each |
Estimated Shipment: 25-04-2025
Log in to see stock. |
||||
17183842
![]() |
Waters Corporation
186006686 |
150 mm | 3 mm | 1.7 μm |
£1309.50
Each |
Estimated Shipment: 25-04-2025
Log in to see stock. |
||||
Description
- The reduction and control of surface silanol activity on Viridis Hybrid particles delivers, under SFC conditions, excellent peak shapes – without the use of mobile-phase additives – even for well-retained basic achiral compounds.
Specifications
6000 psi (415 Bar) | |
130 Å | |
No |
Spherical | |
1/Ea. | |
Column |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Waters Corporation Viridis BEH Column