Learn More
3M™ Virtua™ Safety Glasses, 71500-00001
2.95 GBP valid until 2024-07-31
Use promo code "21674" to get your promotional price.
Use promo code "21674" to get your promotional price.
Click to view available options
Frame Color: Clear
Description
- Unisex safety glasses offering protection and affordability for site visitors or single use protection
- Design provides maximum clarity, excellent coverage and a wide field of vision
- Contoured flexible temples for a close and comfortable fit
- Anti-scratch coating for increased durability
- Clear lens for excellent protection against Ultraviolet (UV) radiation
- Lens marking: 2C-1.2 3M 1 FT
- Frame marking: 3M EN166 FT CE
Specifications
Certifications/Compliance | CE Approved; EN 166:2001 |
Lens Coating | Scratch-resistant |
For Use With (Application) | Offer protection and affordability for site visitors or single-use protection |
Frame Color | Clear |
Temple Style | Flexible |
Lens Material | Polycarbonate |