Learn More
Thermo Scientific™ Virtuoso™ Multiple Vial Guide
Click to view available options
For Use With (Equipment):
Virtuoso Vial Identification System Configured with The Virtuoso Multiple Vial Holder
Unit Size:
Each
Description
Ensure accurate vial identification and sample data integrity by using the Thermo Scientific™ Virtuoso™ Vial Identification System. The system can be configured with the Multiple Vial Holder to hold up to 10 vials and the Multiple Vial Guide to move the vials from the holder to the system for printing one vial at a time in an on-demand fashion.
Specifications
Specifications
| Product Line | Virtuoso |
| Description | Virtuoso Multiple Vial Guide |
| Device Holds | 10 Vials |
| For Use With (Application) | Vial ID and Labelling |
| For Use With (Equipment) | Virtuoso Vial Identification System Configured with The Virtuoso Multiple Vial Holder |