Learn More
Thermo Scientific™ Virtuoso™ Multiple Vial Holder
Print sample data directly on up to 10 vials sequentially for higher throughput using the Thermo Scientific™ Virtuoso™ Multiple Vial Holder.
Brand:  Thermo Scientific™ 60180-VT501
Product Code. 15291918
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Print directly on up to 10 vials sequentially for higher throughput using the Thermo Scientific™ Virtuoso™ Multiple Vial Holder for your Virtuoso Vial Identification System. With advanced printing and labeling capabilities, the Virtuoso system ensures accurate vial identification and the integrity of sample data.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Virtuoso™
10 Vials
Virtuoso Vial Identification System
Virtuoso Multiple Vial Holder
Vial ID and Labelling
5/Pk.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.