missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Virus Transport Medium

Product Code. p-9053026
Click to view available options
Quantity:
1 L
Unit Size:
1000mL
20 L
This item is not returnable. View return policy

Product Code. 16627805

Brand: Gibco™ A48750BA

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A ready-to-use solution containing Hanks balanced salt solution, heat inactivated FBS, gentamicin sulfate, and amphotericin B.

Kit for transporting samples suspected to contain virus

  • It is a ready-to-use solution containing Hanks balanced salt solution, heat inactivated FBS, gentamicin sulfate, and amphotericin B
  • Suitable for the transport of samples suspected to contain virus
  • Kit complies with the formulation published by the Centers for Disease Control and Prevention (CDC) for the preparation of Viral Transport Medium
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 X
Form Liquid
Product Type Virus Transport Medium
Serum Level Standard Serum Supplementation
Sterility Sterile-filtered
Without Additives No Glucose, No Glutamine, No HEPES, No Phenol Red, No Sodium Pyruvate
Quantity 1 L
Shipping Condition Dry Ice
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.