Learn More
SCHOTT AG Lighting and Imaging™ VisiLED Diffuser Only for VisiLED Ring Lights
The VisiLED diffuser comes with improved material and is specially designed only for use with the VisiLED brightfield ring light.
Brand: SCHOTT AG Lighting and Imaging™
Additional Details : Weight : 163.00000kg
Description
VisiLED diffuser made of improved materials is designed specifically for use only with VisiLED brightfield ring lights (S80-65N-2).
- Prevents LED reflections
- Perfect haze of more than 99 % and high transmission
- Includes adapting thread M90 x 1
- Controller needs to be ordered separately
Specifications
Diffuser | |
A Wide Variety of Tasks From Life Science to Industrial Applications | |
VisiLED Ringlight S80-65N-2 |