Learn More
Thermo Scientific™ VISION pro™ UV-Visible Spectrophotometer VISIONpro Software, Single license
Thermo Scientific™ VISION pro™ is full-featured instrument control software for applications from research to routine analysis.
Brand:  Thermo Scientific™ 10040101
Code : 50
Additional Details : Weight : 50.00000kg
Product Code. 10428292
  • £770.00 / Each
Estimated Shipment: 22-08-2024
to see stock.
Description
Description
Thermo Scientific™ VISION pro™ is full-featured instrument control software for applications from research to routine analysis. Powerful, versatile software offers advanced instrument control and data manipulation capabilities. Optimized for Thermo Scientific UV-Visible spectrophotometers, it brings a user-friendly, graphical environment to spectrophotometric analysis. Method setup, data collection, calculations, report presentation, regulatory compliance - all aspects of your analysis are easier with this software.
  • For use with Thermo Scientific Evolution™ 100, 160, 300 and 600 UV-Vis Spectrophotometers
  • Performs Scan, Fixed Wavelength Analysis, Quantification and Multi-Component Analysis
  • Produces spreadsheet results table
  • Arranges data easily with report composer
Includes:
User Manual and RS-232-C cable (Part No. 401317263061)
Requires:
System Requirements: Microsoft™ Windows™ 2000, XP, Vista
Specifications
Specifications
FDA guidelines, IEEE software engineering standards
VISIONpro Software; Single License
Fixed, Scan, Quant and MCA
10040101
Evolution 300
At least Windows XP