Learn More
Thermo Scientific™ VISION security™ UV-Visible Spectrophotometer Software, Single license
Thermo Scientific™ VISION security is designed for laboratories requiring 21 CFR Part 11 compliance.
Brand:  Thermo Scientific™ 10040301
Code : 50
Additional Details : Weight : 20.00000kg
Product Code. 10703894
  • £2725.00 / Each
Estimated Shipment: 22-08-2024
to see stock.
Description
Description
Thermo Scientific™ VISION security is designed for laboratories requiring 21 CFR Part 11 compliance. VISIONsecurity software provides advanced data security and integrity tools allowing you to meet the requirements of the FDA's Title 21, Part 11 requirements for Electronic Records and Electronic Signatures. VISIONsecurity software can be configured on a per user basis, on one system, or across your entire enterprise, allowing easy administration from one user to tens of thousands.
  • Set up users once either on a local computer or a network, no need to configure each computer running each instrument
  • Multiple security features include: same log-in and password used to log into Windows™, customizing each user profile, electronic signatures with full audit trails and traceability
  • For use with Evolution™ 100, 160, 300 and 600 UV-Vis Spectrophotometers
  • Performs Scan, Fixed Wavelength Analysis, Quantification and Multi-Component Analysis
  • Produces spreadsheet-like results table
  • Arranges data easily with report composer
Includes:
User Manual and RS-232-C cable (Part No. 401317263061)
Requires:
System Requirements: Microsoft™ Windows 2000 or XP Professional (All components of VISIONsecurity can also be run on a single NT 4.0/2000/XP PC for stand-alone operation.)
Specifications
Specifications
FDA 21 CFR Part 11 Compliance
QA/QC, Pharmaceutical
Access Control, Electronic Signatures and Audit Trails
VISIONsecurity Software, Single license
Evolution 300
At least Windows XP