Learn More
Thermo Scientific™ VISIONlite™ Spectrophotometer Software
Product Code. p-6137636 Shop All Thermo Scientific Products
Click to view available options
Type:
6-user Software License
VISIONlite 5 Software
VISIONlite 5 Software + Cable
VISIONlite ColorCalc Software
Wine Analysis Software
Unit Size:
Each
Product Code. 12663775
Brand: Thermo Scientific™ 834091300
Thermo Scientific™ VISIONlite™ 5 Software enhances the capabilities of our stand-alone Visible and UV-Visible spectrophotometers. The software enables you use your spectrophotometer to its fullest potential, displaying and reporting experimental and analytical results with ease.
Thermo Scientific™ VISIONlite™ 5 Software enhances the capabilities of our stand-alone Visible and UV-Visible spectrophotometers. The software enables you use your spectrophotometer to its fullest potential, displaying and reporting experimental and analytical results with ease.
Thermo Scientific™ VISIONlite™ ColorCalc software is a powerful stand-alone software package that adds the ability to perform numerous color calculations for a variety of applications. The software records the required transmission or reflectance spectrum and automatically performs calculations using the recorded data.
Thermo Scientific™ VISIONlite™ EnzLab software offers predefined methods and operation procedures for all standard enzymatic test kits including Citric Acid, Lactic Acid, D-Glucose, D-Fructose, Ethanol, Lactose and many more. EnzLab automates and facilitates enzymatic determinations, ensures correct calculations, and provides appropriate analytical reports.
Thermo Scientific™ VISIONlite™ Wine Analysis software allows users to perform an extensive list of predefined photometric, colorimetric and enzymatic analytical procedures common in wine and juice analysis. This PC software features a simple user interface, reliable data recording, and error-free evaluation of results within comprehensive reports.
VISIONlite Software enhancements include:
Scan:
  • Peak pick to identify and label wavelength peaks or valleys
  • Overlay spectra for visual comparison
  • Import, save or export data in native or CSV format for easy archiving and reporting
  • Auto scaling of data plus simple zoom controls for detailed examination of spectra 
Rate:  For SPECTRONIC 200 and GENESYS 30, measure multi-cell kinetics in parallel or serial mode
  • Automatic enzyme activity calculation
  • Automatic data storage plus import or export your data in CSV format
  • Measure up to 100 samples in a single data set 
Fixed:
  • Measure at one or multiple wavelengths
  • Apply factors and equations to your data to generate results directly
  • Addition, subtraction, ratio and baseline correction options
  • Display results with units from a menu or define your own custom unit
  • Run standard bio-tests using the preprogrammed protein and DNA methods
  • Save or print results as a report, or export data in CSV format to use in a template
Quant:  More standards and non-linear curve fitting options offer greater confidence and accuracy than can be accomplished under local control on Thermo Scientific SPECTRONIC 200 and GENESYS 30 Spectrophotometers
  • Choose from 11 pre-programmed reporting units or define your own
  • Apply dilution factors in software to allow you to keep data for all your samples in range without doing off-line calculations
  • Automatic data saving for peace of mind
  • Save or print your results as a report, or export data in CSV format to use in your own template
Performance Verification:
  • Select from the range of available Performance Verification tests that is customized to your instrument’s specifications
  • VISIONlite 5 will prompt you to present the correct reference materials in order, and then print a complete report for your compliance file
Accessory control:
Whether your experiment requires automation to enhance sample throughput or thermostatting to preserve precious biological samples, VISIONlite 5 provides full control of the required accessories.
VISIONlite ColocCalc Software:
  • Full spectrum color scales and data reporting:  Record the required spectra in transmittance or reflectance and calculates color values
  • Application-specific color series: Pt-Co/APHA, whiteness, EP color, wine, etc.
  • Sample thickness transformation allows for the correction for actual sample thickness and front surface reflection effects based upon a single value, the material's refractive index, or a full surface reflectance spectrum
Mathematical calculations and customization
VISIONlite ColorCalc Software has the ability to perform configurable or custom calculations and to store and present your custom-configured methods on the calculations menu.
TRUSTED_SUSTAINABILITY
Specifications
Type VISIONlite 5 Software
For Use With (Equipment) Thermo Scientific SPECTRONIC 200, GENESYS 30, and Evolution 201, 220, 260 Bio Spectrophotometers
Includes DVD with software installer and user manual (in English and German). Display languages: English, German, Spanish, Portuguese, French, Italian, Russian and Korean.
Required Accessories 085-820200 - Standard USB cable for Thermo Scientific SPECTRONIC 200 and GENESYS 30 Spectrophotometers