Learn More
Thermo Scientific™ VisionMate™ ST Single-Tube 2D Barcode Reader
Scan single 2D barcodes instantly into your tracking software or other application with this barcode reader, which connects via USB port.
Brand: Thermo Scientific™ 3125
1480.00 GBP valid until 2024-12-31
Use promo code "23291" to get your promotional price.
Code : 50
Additional Details : Weight : 0.46800kg
Description
Scan 2D barcoded tubes into virtually any application with the compact Thermo Scientific™ VisionMate™ ST Single-Tube 2D Barcode Reader. Plug the unit into an available USB port and allow the drive to automatically install. Hold any Thermo Scientific™ 2D barcoded storage tube over the scanning window and decode the barcode instantly.
- Instant read time
- Simple to install and use
- For individual sample retrieval
- Export via comma-separated variable file, Microsoft™ Excel™, XML, COM port, ODBC or TCP socket
Specifications
VisionMate ST 2-D Bar Code Reader | |
USB | |
Comma-separated variable file, Microsoft™ Excel™ |
Barcode Reader | |
1/Pk., 1/Cs. |