Learn More
Macherey-Nagel™ Visocolor™ Reagent Case for Soil Analysis
Support and maintain healthy, productive and biologically active soil. Macherey-Nagel™ Visocolor™ Reagent Case for Soil Analysis contains all reagents, instruments and accessories needed for economical, fast and convenient preparation of soil extracts for analysis in the field or lab.
Brand: Macherey-Nagel™ 931601
Code : NEW
Additional Details : Weight : 1.00000kg Transport : UN number : 3316 Chem class : 9 Pack group : II
Description
Thorough analysis is a cornerstone to support and maintain healthy soil. To effectively and efficiently plan all measures that affect the soil (fertilization, liming, etc.) it is crucial to determine the important soil parameters first.
- For determination of: Ammonium, Nitrite, Nitrate, Potassium, Phosphate, and pH
- Soil extracts are either prepared with Calcium-Acetate-Lactate solution or CaCl2 solution
- If required, can also be used together with other extraction solutions
- Economical refill reagents can be purchased to replace consumed chemicals:
- Visocolor™ HE Tests: pH 4.0-10.0 and Phosphorus in Soil (reagents/color comparison discs)
- Visocolor™ ECO Tests: Potassium, Ammonium 3, Nitrate, Phosphate
- Quantofix™ Test Strips: Ammonium and Nitrate/Nitrite
- pH-Fix 2.0 – 9.0
- Folded filters MN 616 1/4, 18.5cm
- Pyrophosphate solution
- CaCl2 and CAL stock solutions
- Manual for Visocolor™ Case for Soil Analysis
Specifications
Stocked with all materials for field or lab analysis | |
Visocolor™ | |
110 CaCl2 extractions, 7 CAL extractions, 60–100 tests | |
1 Kit | |
Shelf life of reagents: 18 months |
Soil analysis: determination of Ammonium, Nitrite, Nitrate, Potassium, Phosphate, pH | |
All reagents, instruments and accessories, solid case, manual | |
Pre-packed | |
Varies | |
Multiparameter field kit |