Learn More
R&D Systems™ VisUBlock Mouse on Mouse Blocking Reagent
Blocking reagent for use with primary antibodies from mouse host and mouse tissue.
Brand: R&D Systems™ VB001-01ML
229.60 GBP valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description
- Reduces background staining
- Enables staining of mouse tissue with mouse IgG primary antibodies
Specifications
Store the unopened product at 2-8°C. Do not use past expiration date. | |
1 mL | |
Mouse |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 6
Product Content Correction
Product Title
R&D Systems™ VisUBlock Mouse on Mouse Blocking Reagent > 1 mL
Spot an opportunity for improvement?Share a Content Correction