missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ VisULite ECL Western Blotting Substrate

Ready-to-use ultra high sensitivity Enhanced Chemiluminescent (ECL) substrate.

Brand:  R&D Systems™ VL002-200

Product Code. 30000700

  • £332.00 / 200mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: Substrate A, 100mL; Substrate B, 100mL
This item is not returnable. View return policy

Description

Description

  • High sensitivity – femtogram level detection
  • High signal intensity
  • Excellent signal to noise ratio
  • Compatible with X-ray film or CCD imaging systems
  • Store unopened product at 2° to 8°C. Do not use past expiration date.
Specifications

Specifications

General Laboratory Research Reagent
200 mL (2 x 100 mL)
Store the unopened product at 2°C to 8°C. Do not use past expiration date.
ECL Western Blot Substrate
Substrate A, 100mL, Substrate B, 100mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.