missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Vitamin D2, 97+%
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
Vitamin D2 (ergocalciferol) is a form of vitamin D, a fat-soluble hormone found in food sources
Vitamin D is important for calcium and phosphorus homeostasis, and can play a role in many biological processes including cell growth
Vitamin D2 has a lower binding affinity than Vitamin D3 for vitamin D binding protein
Applications
Research studies show that Vitamin D2 treatment in rats inhibits bladder tumor promotion
Studies show that the treatment of lung carcinoma cells with vitamin D2 in vitro resulted in increased ACE2 protein levels
Vitamin D2 has been seen to interfere with prion protein oligomerization in vitro
Specifications
Specifications
Absorbance | 460 to 490 (265nm, EtOH 96%) |
Melting Point | 115.0°C to 119.0°C |
Color | Beige to White |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.0 to 103% (HPLC) |
Packaging | Glass bottle |
Quantity | 1 g |
Merck Index | 15, 10215 |
Specific Rotation Condition | +82.00 (25.00°C c=3,in acetone) |
Specific Rotation | + 82.00 |
Show More |
Safety and Handling
Product Identifier
- Vitamin D2
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 2
- Specific target organ toxicity Category 1
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H330-Fatal if inhaled.
- H372-Causes damage to organs.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'