missing translation for 'onlineSavingsMsg'
Learn More

Vitamin K{1}

Product Code. 11352866
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11352866

Brand: Thermo Scientific Alfa Aesar L10575.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Important dietary source found in green plants, algae and photosynthetic bacteria. It is used as a vitamin (prothrombogenic) and veterinary antidote for dicoumarol poisoning. It is an essential vitamin for the synthesis of coagulation factors II, VII, IX, and X in the liver. | CAS: 84-80-0 | C31H46O2 | 450.707 g/mol

Vitamin K{1} is an important dietary source found in green plants, algae and photosynthetic bacteria. It is used as a vitamin (prothrombogenic) and veterinary antidote for dicoumarol poisoning. It is an essential vitamin for the synthesis of coagulation factors II, VII, IX, and X in the liver.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84-80-0
Molecular Formula C31H46O2
Molecular Weight (g/mol) 450.707
MDL Number MFCD00214063
InChI Key MBWXNTAXLNYFJB-NKFFZRIASA-N
Synonym vitamin k1, phytonadione, phylloquinone, phytylmenadione, phyllochinon, phytomenadione, alpha-phylloquinone, 3-phytylmenadione, konakion, 2-methyl-3-phytyl-1,4-naphthoquinone
PubChem CID 5284607
ChEBI CHEBI:18067
IUPAC Name 2-methyl-3-[(E,7R,11R)-3,7,11,15-tetramethylhexadec-2-enyl]naphthalene-1,4-dione
SMILES CC1=C(C(=O)C2=CC=CC=C2C1=O)CC=C(C)CCCC(C)CCCC(C)CCCC(C)C

Specifications

Melting Point ∼-20°C
Density 0.984
Boiling Point ∼140°C (0.001mmHg)
Flash Point >110°C (230°F)
Packaging Glass bottle
Refractive Index 1.526
Quantity 1 g
Beilstein 2568816
Sensitivity Light sensitive
Specific Gravity 0.984
Solubility Information Miscible with dehydrated alcohol,acetone,petroleum ether,hexane,dioxane,chloroform,ether,benzene and vegetable oils. Immiscible with water.
Physical Form Viscous Liquid
Chemical Name or Material Vitamin K1
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.