missing translation for 'onlineSavingsMsg'
Learn More

Emma Safety Footwear Vito S3 Shoe

Product Code. p-7225065
Click to view available options
Size:
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
This item is not returnable. View return policy

Product Code. 17572436

Brand: Emma Safety Footwear MM11400039

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Maintain that sharp look, even when wearing your safety gear with these Emma Vito safety shoes.

  • Smart work shoes with water-resistant full-grain and nubuck leather uppers
  • Hydro-Tec antibacterial and Wellness leather lining
  • Antibacterial inlays with leather top layers
  • Steel toecaps and aramid fibre penetration-resistant midsoles
  • Polyurethane (PU) comfort midsoles with thermoplastic polyurethane (TPU) outsoles
  • Regular fit
  • Worn shoes can be returned to the manufacturer for recycling (postage provided); keep receipt and contact us for more details when the time comes
  • Outsoles are resistant to fuel oil
  • Excellent slip resistance
  • Energy-absorbing heels for comfort and support
  • Antistatic

Specifications

Certifications/Compliance ISO 20345:2011 S3 FO
Closure Type Laces
Color Black
For Use With (Application) Government, Local Authority, Oil and Gas
Material Leather
Size 39
Toe Material Steel
Fit Extra-wide Width
Ankle Style Low-cut
Product Type Safety Shoe
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.