Learn More
Aalborg™ Viton A O-ring Stainless Steel Frame with Plug
Aalborg™ Viton A O-ring Stainless Steel Frame with Plug is for use with multiple tube flow meters, which retain most of the unique features associated with single tube units.
Description
- Rib-guided or fluted metering tubes facilitate stable, accurate readings
- Interchangeability of flow tubes and floats
- Manifolding at inlet or outlet
- Ease of installation and exchange of flow tubes
- Chemical compatibility
- Simple means of mounting
Specifications
Frame |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Aalborg™ Viton A O-ring Stainless Steel Frame with Plug