missing translation for 'onlineSavingsMsg'
Learn More

Vitlab™ VITsafe™ Narrow-Mouth Wash Bottles with Imprint

Product Code. p-8000499
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Chemical Label:
Distilled Water
Ethanol
Ethyl Acetate
Unit Size:
Pack of 12
This item is not returnable. View return policy

Product Code. 11857900

Brand: Vitlab™ 1332819

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Identify media quickly and easily with these color-coded wash bottles. VITsafe™ Narrow-Mouth Wash Bottles with Imprint feature a precise spray jet and optimized medium backflow through the continuous and finely drawn tip of the spray tube.

  • Bottles made from LDPE (or PP for Acetone and MEK), with polypropylene spray tube
  • VENT-CAP practically eliminates leakage and prevents almost all static overpressure
  • Color-coded for easy visual identification of the media
Bottles feature durable safety imprint in accordance with Directive (EC) No. 1272/2008 (GHS), as well as with all important information:
  • Material name in German, English, French and Spanish
  • Chemical formula, CAS No., hazard pictogram, signal word
  • Risk phrases (H phrases), safety phrases (P phrases), as well as NFPA Code

Especificaciones

Capacity (Metric) 500 mL
For Use With (Application) Saving and storing
Includes Spray tube and imprint
Material LDPE
No. per Pack 12 Pack
Closure Color Red
Closure Included Yes
Mouth Narrow
Height (Metric) 180 mm
Product Type Wash Bottle
Chemical Label Distilled Water
Closure Material Polypropylene
Diameter (Metric) 74 mm
Closure Type Vented
Mostrar más Mostrar menos
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado