missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VITLAB™ VITsafe™ Wide Mouth Safety Wash Bottles
Click to view available options
Capacity (Metric):
250 mL
500 mL
Chemical Label:
Acetic Acid
Isopropanol
Methanol
Tetrahydrofuran (THF)
Screw Color:
Green
Red
Yellow
Description
Working with chemical substances that are sometimes dangerous requires a high level of responsibility and care. VITsafe™ safety wash-bottles fulfill all safety requirements for safe handling. Spray bottles are individually wrapped in clearly labeled polyethylene bags.
Constructed of durable, transparent materials
Constructed of durable, transparent materials
- Low-density polyethylene (LDPE) bottles
- Spray tubes made with autoclavable, chemically resistant polypropylene (PP)
- Wide-mouth bottles for easy and safe filling, even without a funnel
- Transparent material for easy identification and monitoring
- Material name in German, English, French and Spanish
- Chemical formula, CAS No, hazard pictogram, signal word
- Risk Phrases (H phrases), safety phrases (P phrases), as well as NFPA Code
- In accordance to EC Directive No. 1272/2008 (GHS)
- Design eliminates almost all static overpressure
- Prevents virtually all leaking or dripping that can occur with temperature changes in the lab
- Color-coded caps support easy visual identification of the media
- Bottles utilize screw cap lids with GL threads for a strong, secure hold
- Finely drawn tip creates a precise spray jet for fine control
- Optimized medium backflow through the continuous spray tube
Specifications
Capacity (Metric) | 250 mL |
Color | Transparent |
Material | LDPE |
Packaging | Individually wrapped in PE bag |
Quantity | 12Pack |
Screw Color | Green |
Thread Size (Metric) | GL 45 |
Mouth | Wide |
Height (Metric) | 146 mm |
Chemical Label | Methanol |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'