missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VITLAB™ VITsafe™ 250mL Wide-Mouth Wash Bottles
80.83 GBP valid until 2024-12-31
Use promo code "23277" to get your promotional price.
Use promo code "23277" to get your promotional price.
Click to view available options
Material:
LDPE
Description:
Imprint: Distilled Water
Imprinted: Ethanol
Closure Color:
Blue
Orange
Description
- Manufactured from PE-LD (or PP for Acetone and MEK), spray tube from PP
- Complies in accordance to Directive (EC) No. 1272/2008 (GHS)
- Imprinted with:
- Material name in German, English, French and Spanish
- Chemical formula, CAS No, hazard pictogram, signal word
- Risk Phrases (H phrases), safety phrases (P phrases), as well as NFPA Code
- Precise spray jet and optimised medium backflow through the continuous and finely drawn tip of the spray tube
- The VENT-CAP design oprevents almost all static overpressure minimizing leakage or dripping
Specifications
Capacity (Metric) | 250 mL |
Description | Imprinted: Ethanol |
Material | LDPE |
Closure Color | Orange |
Certifications/Compliance | Directive (EC) No. 1272/2008 (GHS) |
Closure Included | Yes |
Mouth | Wide |
Height (Metric) | 146 mm |
Closure Material | Polypropylene |
Diameter (Metric) | 58 mm |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'