missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Stedim Biotech™ Vivacon 2 125kDa
Centrifugal units for fast DNA concentration and buffer exchange of 2 mL samples.
£205.00 - £735.00
Specifications
Description | MWCO: 125,000 Daltons |
---|---|
Volume (Metric) | 0.1 to 2.0 mL, 55 μL (Dead Stop Volume) |
Product Line | Centrifugal ultrafiltration device |
Material | Polycarbonate Concentrator Body, Polypropylene Filtrate Vessel, Cellulose Acetate Membrane |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15933040
|
Sartorius Stedim Biotech™
VN02H81 |
25/Pk. |
£205.00
Pack of 25 |
In Stock
Log in to see stock. |
|||||
15943040
|
Sartorius Stedim Biotech™
VN02H82 |
100/Pk. |
£735.00
Pack of 100 |
Estimated Shipment: 04-09-2024
Log in to see stock. |
|||||
Description
- The product of choice for protein fractionation applications.
- The use of Hydrosart™ membranes results in high recoveries from dilute samples.
- Complete and highly reproducible recovery of the concentrate.
- Reverse spinning step capacity.
- Concentration of up to 0.5 mL genomic DNA or peptide samples.
- Low binding material.
- MWCO printed on device.
- Selected cut offs available sterile and DNA-free for things like forensic applications.
- qPCR assay over 55 cycles with a verified limit of detection < 2 pg
Specifications
MWCO: 125,000 Daltons | |
0.1 to 2.0 mL, 55 μL (Dead Stop Volume) | |
Centrifugal ultrafiltration device | |
Polycarbonate Concentrator Body, Polypropylene Filtrate Vessel, Cellulose Acetate Membrane |