missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Stedim Biotech™ Vivacon 500 125kDa
Vivacon 500 125kDa are centrifugal units for fast DNA concentration and buffer exchange of 500 μL samples.
£169.00 - £560.00
Specifications
Volume (Metric) | 0.1 to 0.5 mL, 5 μL (Dead Stop Volume) |
---|---|
Product Line | Centrifugal ultrafiltration device |
Sterility | Sterile |
Material | Polycarbonate Concentrator Body, Polypropylene Filtrate Vessel, Cellulose Acetate Membrane |
Closure Included | Yes |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15913040
|
Sartorius Stedim Biotech™
VN01H81 |
25 Pk. |
£169.00
Pack of 25 |
In Stock
Log in to see stock. |
|||||
15923040
|
Sartorius Stedim Biotech™
VN01H82 |
100 Pk. |
£560.00
Pack of 100 |
In Stock
Log in to see stock. |
|||||
Description
- The product of choice for protein fractionation applications.
- The use of Hydrosart™ membranes results in high recoveries from dilute samples.
- Complete and highly reproducible recovery of the concentrate.
- Reverse spinning step capacity.
- Concentration of up to 0.5 mL genomic DNA or peptide samples.
- Low binding material.
- MWCO printed on device.
- Selected cut offs available sterile and DNA-free for things like forensic applications.
- qPCR assay over 55 cycles with a verified limit of detection <2 pg
Specifications
0.1 to 0.5 mL, 5 μL (Dead Stop Volume) | |
Sterile | |
Yes | |
Centrifugal Units |
Centrifugal ultrafiltration device | |
Polycarbonate Concentrator Body, Polypropylene Filtrate Vessel, Cellulose Acetate Membrane | |
Single-use |