missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Vivacon™ 500 DNA Concentrator
BIOPHARMA

Product Code. 11716355 Shop All Sartorius Products
Click to view available options
MWCO:
10 kDa
100 kDa
2 kDa
30 kDa
50 kDa
Quantity:
100 Pack
25 Pack
Unit Size:
Pack of 100
Pack of 25
This item is not returnable. View return policy

Product Code. 11716355

Brand: Sartorius VN01H92

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For up to 500 μL, Vivacon™ 500 are centrifugal units for fast DNA concentration and buffer exchange of 500 μL samples. They are also the product of choice for protein fractionation applications. The use of Hydrosart™ membranes results in high recoveries from dilute samples.

Vivacon 500 are centrifugal devices for DNA concentration and buffer exchange applications for samples up to 500μl. They are also the product of choice for protein fractionation applications in this volume range. For optimal performance with very diluted samples, e.g. forensic specimens, Vivacon 500 is equipped with patented Hydrosart™ regenerated cellulose membranes with extremely low unspecific binding properties and high flux. High recoveries and excellent reproducibility are paired with convenience offered by volume graduation and molecular weight cut-offs printed on individual devices. Vivacon 500 features a horizontal membrane design optimal for DNA fractionation (primer removal) as well as protein fractionation (peptide removal). The possibility of a reverse spin after sample processing assures complete concentrate recovery which is especially important when working with low sample concentrations.

  • Convenient handling in fixed-angle rotors
  • Low binding material
  • Reverse spin possibility
  • MWCO printed on
  • Complete and highly reproducible concentrate recovery
TRUSTED_SUSTAINABILITY

Specifications

Concentration 50ng/mL
Concentration Time 60 min.
Filter Type Ultrafiltration
For Use With (Equipment) Centrifuge
Material Polycarbonate
Material (Membrane) Regenerated Cellulose
MWCO 2 kDa
Sample Volume 500 μL
Sterility Non-sterile
Filtration Area 0.32 cm2
For Use With (Application) DNA concentration, desalting, buffer exchange, fractionation
Max. Force 7500xG
Quantity 100 Pack
Sample Type DNA, peptide
Type Concentrator
Volume (Metric) 0.5 mL
Volume (Metric) Hold Up <5 μL
Dimensions (L x W) 1.77 x 0.48 in. (4.5 x 1.24cm)
Length (English) 1.77 in.
Length (English) Back Spin 1.87 in.
Length (Metric) 4.5 cm
Length (Metric) Back Spin 4.75 cm
Width (English) 0.48 in.
Width (Metric) 1.24 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.