Learn More
Sartorius™ Vivacon™ Polycarbonate/Polypropylene Centrifugal Unit
Brand:  Sartorius™ VN01H21ETO
 View more versions of this product
Product Code. 11735785
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Specifications
PCR
30 kDa
Polycarbonate, Polypropylene
DNA
0.1 to 0.5 mL
Polycarbonate, Polypropylene
Concentration, Desalting, buffer exchange
25/Pk.
Vivacon 500 Centrifugal Unit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Sartorius™ Vivacon™ Polycarbonate/Polypropylene Centrifugal Unit > Volume: 0.1 to 0.5mL; Molecular Weight: 30,000dalton; Quantity: 25/pk