Learn More
Sartorius™ Vivacon™ Polycarbonate/Polypropylene Centrifugal Unit
Brand: Sartorius™ VN01H22ETO
This item is not returnable.
View return policy
Description
Specifications
PCR | |
30,000 Da | |
Polycarbonate, Polypropylene | |
DNA | |
0.1 to 0.5 mL |
Polycarbonate, Polypropylene | |
Concentration, Desalting, buffer exchange | |
100/Pk. | |
Vivacon 500 Centrifugal Unit |