Learn More
Sartorius™ Vivaflow™ 50R, 200 Accessory Set
Complete set of accessories for Vivaflow 50R, 200 with 230V pump.
Brand:  Sartorius™ VFS202
Code : NEW
Additional Details : Weight : 0.01000kg
Product Code. 11755046
  • £3025.00 / Each
Estimated Shipment: 05-09-2024
to see stock.
Add to basket
Includes: pump, easy-load pump head, tubing, diafiltration reservoir, module stand, pressure indicator, T-connectors, series interconnectors
Description
Description
  • Includes pump, easy-load pump head, tubing, diafiltration reservoir, module stand, pressure indicator, T-connectors, series interconnectors
Specifications
Specifications
Vivaflow 50R, 200 Accessory Set