Learn More
Sartorius Vivapore 10-20 Expansion Reservoir
Vivapore 10-20 expansion reservoir with simple push-fit handling is compatible with Vivapore 10 solvent absorption concentrators. It can either be used to increase the sample capacity of Vivapore from 10 to 20 mL, or for continuous diafiltration for sample desalting and buffer exchange applications.
Brand: Sartorius VP-ARV0010-D
Additional Details : Weight : 0.09000kg
Description
- Expand or exchange
- Optional reservoirs
- Flexibility to double sample capacity
- Concentrate and desalt in one step
Specifications
Polypropylene | |
Non-sterile | |
Reservoir | |
10 mL |