Learn More
Sartorius Vivapore 5
Vivapore 5 solvent absorption concentrators are disposable ultrafilters for macromolecule concentration from up to 5 mL aqueous samples.
Brand: Sartorius VP-S005P0008-3
Additional Details : Weight : 0.65000kg
Description
- Concentrate equipment-free
- Fill and go
- Tailored process monitoring
- Maintained sample integrity
- Sensitive molecule friendly
Specifications
Ultrafiltration | |
PES | |
5 mL | |
Solvent Absorption Concentrator |
SAN | |
7.5 kDa | |
Non-sterile |