Learn More
Sartorius Vivapore Stand
Vivapore stand can support up to four Vivapore 5 or 10 solvent absorption concentrators during use with simple push-fit handling.
Brand: Sartorius VP-AST0001-C
Additional Details : Weight : 0.20000kg
Description
- Simple push-fit handling
- Stable Support
Specifications
ABS | |
Non-sterile | |
Stand |