Learn More
Cytiva Vivaspin™ 2 Sample Concentrators
Use for one-step, non-denaturing sample concentration in a single tube for minimal sample handling and reduced sample loss. Cytiva Life Vivaspin™ 2 Sample Concentrators cater to scientists working with sample volumes ranging from 100 μl to 20 ml, with a range of MWCO values from Mr 3 000 to 100 000.
£165.00
Specifications
Material (Membrane) | PES |
---|---|
Filtration Area | 1.2 cm2 |
Quantity | 25/Pk. |
Volume (Metric) | 400 μL to 2 mL |
Length (Metric) | 126 mm |
Product Code | Brand | MWCO | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | MWCO | Price | Quantity & Availability | |||||
10023147
|
Cytiva
28-9322-48 |
30 kDa |
£165.00
Pack of 25 |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||||
10723837
|
Cytiva
28-9322-45 |
5 kDa |
£165.00
Pack of 25 |
Estimated Shipment: 15-07-2024
Log in to see stock. |
|||||
10259714
|
Cytiva
28-9322-57 |
50 kDa |
£165.00
Pack of 25 |
Estimated Shipment: 16-07-2024
Log in to see stock. |
|||||
10724027
|
Cytiva
28-9322-47 |
10 kDa |
£165.00
Pack of 25 |
Estimated Shipment: 16-07-2024
Log in to see stock. |
|||||
10157244
|
Cytiva
28-9322-40 |
3 kDa |
£165.00
Pack of 25 |
Estimated Shipment: 16-07-2024
Log in to see stock. |
|||||
10269714
|
Cytiva
28-9322-58 |
100 kDa |
£165.00
Pack of 25 |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||||
Description
Up to 30-fold concentration of the sample can be achieved with recovery of the target molecule typically exceeding 95%. Patented dead-stop technology ensures that samples cannot be concentrated to dryness and enables direct concentrate recovery.
- Accept sample volumes from 100μLto 20mL, with a range of molecular weight cutoff values from Mr3000 to 100,000
- Vertical polyethersulfone semi-permeable membrane separating upper and lower chamber resists blockage and tolerates high flow rate
- Centrifugation forces solvent through membrane, leaving concentrated sample in the upper chamber
- Compatible pH range: 1 to 9
- Active membrane area: 1.2cm2
- Vivaspin 2 offers easy, contact-free storage by reverse spinning the concentrate into the recovery cap
For maximum recovery, select a MWCO at least 50% smaller than the molecular size of the species of interest.
Specifications
PES | |
25/Pk. | |
126 mm |
1.2 cm2 | |
400 μL to 2 mL | |
17 mm |