missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Vivaspin™ 20 Centrifugal Concentrator
BIOPHARMA

Offers increased volume flexibility and performance. Vivaspin™ 20 Centrifugal Concentrator handles up to 20mL in swing bucket rotors and 14mL in fixed angle rotors accepting 50mL centrifuge tubes. Features twin vertical membranes for unparalleled filtration speed and >100-fold concentration.

Brand:  Sartorius VS2031

173.92 GBP valid until 2024-12-31
Use promo code "21930" to get your promotional price.


 View more versions of this product

Product Code. 10273521

  • £215.00 / Pack of 12
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Offers increased volume flexibility and performance. VivaspinTM 20 Centrifugal Concentrator handles up to 20mL in swing bucket rotors and 14mL in fixed angle rotors accepting 50mL centrifuge tubes. Features twin vertical membranes for unparalleled filtration speed and >100-fold concentration.

Vivaspin™ 20 centrifugal concentrators have been developed to offer increased volume flexibility and performance. Vivaspin™ 20 handles up to 20mL in swing bucket rotors and 14mL in fixed angle rotors accepting 50mL centrifuge tubes. Vivaspin™ 20 features twin vertical membranes for unparalleled filtration speed and >100-fold concentration. Volume graduation allows to monitor the concentration process and the modified dead stop pocket simplifies pipetting of the final concentrate. For higher process flexibility, Vivaspin™ 20 is available with unique accessories and operating methods. For one-step desalting following concentration, a diafiltration cup filled with buffer is placed on the concentrator and spun to achieve 98% salt removal, instead of two spins to achieve the same result with the traditional refill and re-spin procedure. When an appropriate centrifuge is unavailable, or for single sample processing, for gas pressure filtration, Vivaspin™ 20 can be filled with up to 15ml and then pressurised for bench top concentration. For even faster processing, gas pressure filtration can be combined with centrifugation. Pressure-fugation is particularly useful for difficult or viscous samples such as serum, when using a low process temperature, or generally when minimum process time is essential.

  • High flux PES membrane
  • High retentate recovery > 90 %
  • Vertical twin membrane for ultra fast concentration
  • Universal rotor compatibility
  • Safe sample concentration due to dead-stop volume
  • Convenient concentrate recovery by pipetting
  • Volume graduation for easy reading/monitoring
  • 100-fold concentration with twin vertical membranes
  • Additional option for pressurized operation

Rotor Compatibility
Please note: Vivaspin™ 20 (30mm × 116mm) is designed to fit into rotors that can accommodate Falcon 50 ml conical bottom tubes, e.g. Beckman Allegra 25R with TS-5.1-500 swing-out rotor with BUC 5 buckets and 368327 adaptors; Beckman TA-10.250 25° fixed angle rotor with 356966 adaptors; Multifuge 3 S-R with 75006445 swing out rotor with 75006441 buckets and adaptors for Falcon 50 ml conical bottom tubes. These devices are not designed to fit into rotors that only accept round bottom 29mm × 105mm tubes, e.g. Sorvall SS34 or Beckmann JA 20. If your rotor accepts only 29mm × 105mm round bottom tubes, please use the Vivaspin™ 15, which can be used in either round bottom or conical centrifuge tubes

TRUSTED_SUSTAINABILITY
Specifications

Specifications

PES
12/Pk.
116 mm
50 kDa
14 mL (Fixed Angle Rotor), 20 mL (Swing Bucket Rotor)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.